Skip to Content
Merck
All Photos(1)

Key Documents

CDE-509

Sigma-Aldrich

Diether Liposomes

14:0 Diether PC:Cholesterol (70:30 molar ratio 1mM)

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$993.00

$993.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
2 ML
$993.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

$993.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

composition

Phosphate buffered saline

concentration

1 mM

impurities

30 mol % Cholesterol
70 mol % 14:0 Diether PC

particle size

100 nm

pH

7.4

Looking for similar products? Visit Product Comparison Guide

General description

Liposomes are extensively used to study the interaction of proteins, peptides and other molecules with the surface of a lipid membrane. Ether lipids are a unique class of glycerophospholipids that have an alkyl chain attached to the sn-1 position by an ether bond.

Due to distinctive properties of ether lipids, liposomes made from ether lipids exhibit very unique characters and performance:
*ether bonds are more stable than ester linkages over a wide range of acidic or alkaline pH
*stability properties of the liposomes are enhanced by biopolar lipids and the saturated alkyl chains gives stability towards degradation in oxidative conditions
*the unusual stereochemistry of the glycerol backbone enhances the resistance against phopholipases

Due to their increased stability properties, ether liposomes are suitable candates for experiments that require an extended period of time as they can neither be hydrolyzed, oxidized, and can withstand higher temperatures.

Application

Drug delivery
Lipid-protein interactions

Storage and Stability

Liposomes should never be frozen. Liposomes should be stored in the dark at 4°C, except when brought to room temperature for brief periods prior to use.

Liposomes are made under sterile conditions. If you need to take multiple aliquots out of the vial, it is advised to take extreme care in not contaminating the vial. It is recommended to handle the vial under a sterile hood to maintain the sterility of the product. Liposomes should never be frozen. Ice crystals that form during freezing will rupture the lipid membrane of the liposomes and change the size of liposomes particles.

Legal Information

Cellsome is a trademark of Encapsula NanoSciences
Product of Encapsula Nanosciences

Disclaimer

For research use only

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service