Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diether Liposomes
Products
Brand
Sigma-Aldrich (9)
Chemical Composition
Quality Segment
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
CDE-507
Diether Liposomes,
12:0 Diether PC:Cholesterol (70:30 molar ratio 1mM)
Expand
View Pricing
CDE-504
Diether Liposomes,
100% 16:0 Diether PC (1mM)
Expand
View Pricing
CDE-510
Diether Liposomes,
16:0 Diether PC:Cholesterol (70:30 molar ratio 1mM)
Expand
View Pricing
CDE-506
Diether Liposomes,
100% 18:1 Diether PC (1mM)
Expand
View Pricing
CDE-508
Diether Liposomes,
13:0 Diether PC:Cholesterol (70:30 molar ratio 1mM)
Expand
View Pricing
CDE-512
Diether Liposomes,
18:1 Diether PC:Cholesterol (70:30 molar ratio 1mM)
Expand
View Pricing
CDE-505
Diether Liposomes,
100% 18:0 Diether PC (1mM)
Expand
View Pricing
CDE-509
Diether Liposomes,
14:0 Diether PC:Cholesterol (70:30 molar ratio 1mM)
Expand
View Pricing
CDE-511
Diether Liposomes,
18:0 Diether PC:Cholesterol (70:30 molar ratio 1mM)
Expand
View Pricing