Skip to Content
Merck
All Photos(1)

Key Documents

C4080

Sigma-Aldrich

Cyheptamide

Synonym(s):

10,11-Dihydro-5H-dibenzo(a,d)cycloheptene-5-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$84.30

$84.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$84.30

About This Item

Empirical Formula (Hill Notation):
C16H15NO
CAS Number:
Molecular Weight:
237.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.30


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

SMILES string

NC(=O)C1c2ccccc2CCc3ccccc13

InChI

1S/C16H15NO/c17-16(18)15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8,15H,9-10H2,(H2,17,18)

InChI key

APBVLLORZMAWKI-UHFFFAOYSA-N

Application

Used to study pharmacological activity involved in identification of compounds with central nervous system activity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anticonvulsant activities and brain concentrations of cyheptamide and carbamazepine.
G H Wimbish et al.
Proceedings of the Western Pharmacology Society, 23, 75-79 (1980-01-01)
Josefin Koehn et al.
Antimicrobial agents and chemotherapy, 59(11), 6682-6688 (2015-08-08)
In the treatment of HIV infection, a combination of anti-HIV drugs is commonly used in highly active antiretroviral therapy (HAART). One such combination recommended for clinical therapy consists of the two HIV protease inhibitors atazanavir and ritonavir and the HIV
G L Jones et al.
Journal of pharmaceutical sciences, 70(6), 618-620 (1981-06-01)
Carbamazepine and cyheptamide have certain stereochemical features in common with phenytoin; when superimposed, two bulky hydrophobic groups in each permit the approximate apposition of two electron donor atoms. The anticonvulsant activity of each compound was determined in mice using a
P W Codding et al.
Journal of medicinal chemistry, 27(5), 649-654 (1984-05-01)
The molecular structures of cyheptamide and 3-hydroxy-3- phenacyloxindole were determined by X-ray diffraction methods. The amide group in both compounds exhibits delocalization of the pi-electrons over the three atoms (N, C, and O), while the bond linking the amide to
[Determination of plasma lidocaine by gas-liquid chromatography].
I Saavedra et al.
Revista medica de Chile, 115(7), 661-664 (1987-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service