Skip to Content
Merck
All Photos(2)

Key Documents

D31737

Sigma-Aldrich

5-Dibenzosuberenone

97%

Synonym(s):

5H-Dibenzo[a,d]cyclohepten-5-one

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$157.00
100 G
$284.00

$157.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$157.00
100 G
$284.00

About This Item

Empirical Formula (Hill Notation):
C15H10O
CAS Number:
Molecular Weight:
206.24
Beilstein:
743171
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$157.00


Available to ship on17 April 2025Details


Request a Bulk Order

Assay

97%

form

powder

mp

87-88 °C (lit.)

SMILES string

O=C1c2ccccc2C=Cc3ccccc13

InChI

1S/C15H10O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-10H

InChI key

SNVTZAIYUGUKNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service