Skip to Content
Merck
All Photos(1)

Key Documents

BR781702

BRAND® microplate BRANDplates®, pureGrade

size 1536 wells, size 10 μL, flat bottom, black

Synonym(s):

BRANDplates®, Microplates, pureGradeS

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$2,450.00

$2,450.00


Estimated to ship on12 April 2025



Select a Size

Change View
50 EA
$2,450.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
black wells
flat bottom
polystyrene
size:
10 μL
1536 wells
working volume:
10 μL

$2,450.00


Estimated to ship on12 April 2025


material

black wells
flat bottom
polystyrene

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 50 ea
pkg of (individually packaged)

manufacturer/tradename

BRAND 781702

size

10 μL
1536 wells

working volume

10 μL

color

black

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service