Skip to Content
Merck
All Photos(1)

Key Documents

P0674

Sigma-Aldrich

Microcapillary tube, calibrated

capacity 10 μL

Synonym(s):

Calibrated Capillary Tube, Micropipette Tube

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$165.00

$165.00


Available to ship on11 April 2025Details



Select a Size

Change View
1 PKG
$165.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

$165.00


Available to ship on11 April 2025Details


material

glass

packaging

pack of 250 ea

manufacturer/tradename

Drummond Scientific 2-000-010

measuring range

accuracy: ±0.5%

capacity

10 μL

color

color-coded orange

Looking for similar products? Visit Product Comparison Guide

General description

  • Drummond Calibrated Micropipettes provide accuracy and convenience for a wide variety of liquid transfer applications.
  • Made of high quality glass.
  • Fire polished micropipettes are imprinted with volume and a color-code bar for easy identification.
  • Calibrated "To Deliver."
High quality Drummond micropipettes are fire polished, imprinted with volume and a color-code bar for easy identification. Calibrated "To Deliver."

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William T Keenan et al.
Frontiers in neural circuits, 11, 93-93 (2017-12-09)
Designer Receptors Exclusively Activated by Designer Drugs (DREADDs) are an important tool for modulating and understanding neural circuits. Depending on the DREADD system used, DREADD-targeted neurons can be activated or repressed
Rachel Posner et al.
Cell, 177(7), 1814-1826 (2019-06-11)
It is unknown whether the activity of the nervous system can be inherited. In Caenorhabditis elegans nematodes, parental responses can transmit heritable small RNAs that regulate gene expression transgenerationally. In this study, we show that a neuronal process can impact
Elisabet Rosàs-Canyelles et al.
Science advances, 6(17), eaay1751-eaay1751 (2020-06-05)
The process by which a zygote develops from a single cell into a multicellular organism is poorly understood. Advances are hindered by detection specificity and sensitivity limitations of single-cell protein tools and by challenges in integrating multimodal data. We introduce

Questions

  1. What is the diameter of the outside of the capillary?

    1 answer
    1. The diameter of the capillary tubes is as follows:
      O.D. - 1.041 mm
      I.D. - 0.5011 mm

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service