BR74506
BRAND® filter funnel support with base plate
for 2 funnels
Synonym(s):
Accessories, Filter funnels, Funnels, Supports
All Photos(1)
About This Item
Recommended Products
material
polypropylene
feature
for 2 funnels
manufacturer/tradename
BRAND 74506
L × W
250 mm × 140 mm
General description
These PP plates feature a stainless steel support rod with dimensions of 12 mm in diameter and 595 mm in length. They are designed to support 2 or 4 funnels with a top outer diameter ranging from 50 mm to 120 mm. The height of the funnels is adjustable, allowing for customized setups based on your specific needs.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG