Key Documents
BR701381
BRAND® Sealing Paddle
Select a Size
1 EA
$15.30
$15.30
Available to ship on23 April 2025Details
Select a Size
Change View
1 EA
$15.30
About This Item
UNSPSC Code:
41122410
NACRES:
NB.13
$15.30
Available to ship on23 April 2025Details
material
polyethylene
sterility
non-sterile
manufacturer/tradename
BRAND 701381
Related Categories
General description
To securely and easily apply self-adhesive films. The sealing paddle helps to attach the film to the plate with a uniform pressure. Especially suited for plates with raised rim.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)