Key Documents
BR472000
BRAND® staining tray for glass staining trough
soda-lime glass
Synonym(s):
Microscopy, Staining, Staining Accessories
Select a Size
10 EA
$421.00
$421.00
Available to ship on23 April 2025Details
Select a Size
Change View
10 EA
$421.00
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
$421.00
Available to ship on23 April 2025Details
material
soda-lime glass
description
use with glass staining trough
manufacturer/tradename
BRAND 472000
L × W × H
88 mm × 70 mm × 40 mm, holds 10 slides
General description
staining trough, tray, and wire handle are sold separately
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)