Key Documents
BR43338
BRAND® BLAUBRAND® density bottle, Gay-Lussac pattern, calibrated
capacity 100 mL, borosilicate glass
Synonym(s):
BLAUBRAND®, Gay-Lussac
Select a Size
1 EA
$182.00
$182.00
Available to ship on14 April 2025Details
Select a Size
Change View
1 EA
$182.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.71
$182.00
Available to ship on14 April 2025Details
material
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Joint
joint ST/NS neck10/19
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 43338
capacity
100 mL
General description
DIN ISO 3507, type Gay-Lussac. Calibrated to contain (TC, In). Incl. individual certificate. Stopper NS 10/19 with capillary. Top of stopper ground and polished. The volume in cm³ is specified to a precision of 3 decimal places.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.