BR22316
BRAND® BLAUBRAND® automatic burette, Pellet pattern
volume 25 mL, accuracy: ±0.08 mL, amber borosilicate glass (lateral needle-valve stopcock PTFE-key, grad. in white)
Synonym(s):
Automatic Burettes, Burettes, Pellet pattern, SILBERBRAND
About This Item
Recommended Products
material
amber borosilicate glass (lateral needle-valve stopcock PTFE-key, grad. in white)
feature
subdivision 0.1 mL
SILBERBRAND class B
without bottle
without intermediate stopcock
manufacturer/tradename
BRAND 22316
volume measuring range
25 mL, accuracy: ±0.08 mL
volume
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
All BLAUBRAND burettes and automatic burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service