BR137634
BRAND® magnetic stirring bar, PTFE, crosshead
crosshead, H × D 10 mm × 25 mm
Synonym(s):
Magnetic Stirring Bars, PTFE
Select a Size
$168.00
Available to ship on23 April 2025Details
Select a Size
About This Item
$168.00
Available to ship on23 April 2025Details
Recommended Products
material
PTFE-coated
crosshead
manufacturer/tradename
BRAND 137634
H × D
10 mm × 25 mm
L
25 mm
diam.
10 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service