Skip to Content
Merck
All Photos(1)

Key Documents

BMSE3100E

Benchmark Accuris MyView Compact UV Transilluminator

AC/DC input 230 V, EuroPlug

Synonym(s):

ethidium bromide, gel imaging, gel casting, transillumination

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,350.00

$1,350.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$1,350.00

About This Item

UNSPSC Code:
41106000
NACRES:
NC.01

$1,350.00


Check Cart for Availability

manufacturer/tradename

Benchmark Scientific E3100-E

Looking for similar products? Visit Product Comparison Guide

General description

The myView Transilluminator has a surprisingly small footprint (smaller than a standard piece of paper). However, the viewing surface is large enough to accommodate a midsized gel or multiple smaller gels. The 16.5 x 13.5 cm filter glass is scratch resistant to allow band excision with a metal blade. The four 6W bulbs provide and evenly illuminated surface for viewing gels and capturing quality images.

Application

Electrophoresis

Legal Information

Accuris is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service