Skip to Content
Merck
All Photos(1)

Key Documents

BMSE4000E

Benchmark SmartBlue Transilluminator

AC/DC input 230 V, EuroPlug

Synonym(s):

SYBR, SmartGlow, blue light, gel imaging, gel casting, transillumination

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,120.00

$1,120.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$1,120.00

About This Item

UNSPSC Code:
41106000
NACRES:
NB.13

$1,120.00


Check Cart for Availability

manufacturer/tradename

Benchmark Scientific E4000-E

Looking for similar products? Visit Product Comparison Guide

General description

Accuris SmartBlue Transilluminators have been developed for viewing DNA gels stained with new, safe fluorescent dyes. These new dyes, such as SmartGlow, SYBR Green, Gel Green, have an excitation wavelength of 465470nm (visible blue light), and are safer to use than Ethidium Bromide.

Application

Electrophoresis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service