Key Documents
BAF379001100
SP Bel-Art® Minipet Aqueous Pipettor
capacity 10 mL
Synonym(s):
Pipette
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F37900-1100
capacity
10 mL
General description
Minipet Pipettors feature a three-way check valve for continuous, repetitive pipetting. Just place the filling line in a reservoir, push the PTFE® plunger to dispense and release to refill.
Features and Benefits
  • Molded polypropylene handle offers comfort for extended use
    Turning the knob on top sets the self-locking capacity to assure dispensing repeatability of ± 0.5%; extensions can be added to the filling line for greater reach
    PTFE® Minipet Pipettors offer a high level of chemical resistance; may be used with virtually any solvent or acid (except HF); fluids come in contact with only borosilicate glass and PTFE®; Supplied with an all-PTFE® valve (with glass check) and PTFE® sinker on 45.7cm (18”) line, 2.4mm I.D.(⅛”)
    Aqueous Minipet Pipettors are autoclavable and can be used with a Luer lock manifold for multi-well plate work; supplied with a chrome plated check valve, Luer lock fitting, Buna-N-seals, stainless steel spring, and Luer lock cannula, 45.7cm long (18”), 3.2mm (⅛”) I.D. silicon filling tube and PTFE® sinker
    PTFE® is a registered trademark of The Chemours Company FC LLC and used under license by Bel-Art Products, Inc.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company