Key Documents
BAF378970000
SP Bel-Art® Pipette Pump
capacity 2 mL, blue
Synonym(s):
Pump For Pipette, pipetting device
Select a Size
1 EACH
$54.10
$54.10
Estimated to ship on12 April 2025
Select a Size
Change View
1 EACH
$54.10
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
$54.10
Estimated to ship on12 April 2025
material
blue
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F37897-0000
capacity
2 mL
General description
Provides safe, accurate and trouble-free pipetting with simple, one-handed operation.
Features and Benefits
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)