BAF187990000
Scienceware® test tube capper
Polypropylene with vinyl covered handhold
Select a Size
$103.00
Select a Size
About This Item
$103.00
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18799-0000
diam. × H
4.4 cm × 8.9 cm
General description
- Capping can be performed with test tubes in a rack or while being held in the hand
- Designed for use with recessed center test tube caps
- Polypropylene with vinyl covered handhold
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)