Skip to Content
Merck
All Photos(2)

Key Documents

BAF170940020

Scienceware® Aquet® liquid detergent

volume 20 x 20 mL , pouches

Synonym(s):

liquid detergent

Sign Into View Organizational & Contract Pricing

Select a Size

20 EACH
$123.00

$123.00


Check Cart for Availability


Select a Size

Change View
20 EACH
$123.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

$123.00


Check Cart for Availability

packaging

pack of 20 ea

manufacturer/tradename

Bel-Art F17094-0020

volume

20 x 20 mL , pouches

General description

Safe for glassware, plastics and surfaces, this phosphate-free, nonionic surfactant will not etch or mar precision glassware. It rinses quickly and dries without wiping.

Legal Information

Aquet is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service