Skip to Content
Merck
All Photos(2)

Key Documents

BAF131643138

Scienceware® biohazard disposal bags

width × H × thickness 790 mm (31 in.) × 38 in. (970 mm) × 0.04 mm (1.5 mil), pack of 200 ea, red

Synonym(s):

Scienceware® biohazard disposal bags

Sign Into View Organizational & Contract Pricing

Select a Size

200 EACH
$906.00

$906.00


Check Cart for Availability


Select a Size

Change View
200 EACH
$906.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.35

$906.00


Check Cart for Availability

material

red

packaging

pack of 200 ea

manufacturer/tradename

Bel-Art F13164-3138

capacity

49.2-75.7 L , metric

width × H × thickness

790 mm (31 in.) × 38 in. (970 mm) × 0.04 mm (1.5 mil)

Looking for similar products? Visit Product Comparison Guide

General description

Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.

Features and Benefits

Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • High temperature steam autoclavable to 135°C (275°F)
  • Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
  • Made in USA

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service