Key Documents
BAF118470010
Polyethylene carboy with spigot
capacity 4 L (1 gal), polyethylene
Synonym(s):
Polyethylene carboy with spigot
Select a Size
1 EACH
$325.00
$325.00
Check Cart for Availability
Select a Size
Change View
1 EACH
$325.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
$325.00
Check Cart for Availability
material
polyethylene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F11847-0010
capacity
4 L (1 gal)
General description
These bottles and carboys have a screw in dispensing spigot that can be replaced with a plug (included) when the vessels are used for long-term storage. Heavy knurling on the cap provides excellent grip for opening and closing. The smooth sides accept adhesive labels and can be marked with most indelible markers.
  • Spigot opens fully in one half-turn
  • Molded of polyethylene, which resists corrosive acids and alkalis
  • Extra heavy walls, yet lighter than glass
Use to dispense cleaning solutions, disinfectants, laboratory reagents, and intermediate phase mixtures and slurries for various manufacturing processes.
Features and Benefits
  • Use to dispense cleaning solutions, disinfectants, laboratory reagents, and intermediate phase mixtures and slurries for various manufacturing processes.
  • Lightweight, durable polyethylene with extra-heavy walls for long life and dependability
  • Lighter than glass and easy to handle
  • Heavy knurling on cap provides excellent grip for opening and closing
  • Resistant to corrosive acids and alkalis
  • The spigot requires only half turn to open or close
  • Smooth sides accept adhesive labels and can be marked with most indelible markers
Choose from one of the most recent versions:
Certificates of Analysis (COA)