Skip to Content
Merck
All Photos(1)

Key Documents

BAB605100100

SP Bel-Art® H-B Enviro-Safe® Dry Block/Incubator Liquid-In-Glass Thermometer

temp 24-57 °C, PFA (coated), immersion level, 35 mm Partial Immersion: (1.4”)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$179.00

$179.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$179.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.79

$179.00


Check Cart for Availability

material

PFA (coated)

feature

graduation 0.5 °C
immersion level 35 mm Partial Immersion: (1.4”)

packaging

pack of 1 ea

manufacturer/tradename

HB Instruments B60510-0100

temp measuring range

24-57 °C
accuracy: 0.5 °C

Looking for similar products? Visit Product Comparison Guide

General description

These 175mm (6.9”) long, USA made Enviro-Safe® thermometers are EnviroKleen certified environmentally friendly and features 100% non-toxic, non-hazardous, biodegradable, Enviro-Safe® green liquid against white back glass, and recyclable Safety-Pak packaging. Enviro-Safe® thermometers have the same accuracy as similar mercury thermometers, but without the hazards. Item B60510-0100 features PFA flouropolymer safety coating to keep liquid and glass encapsulated in the event of breakage.

Features and Benefits

These 175mm (6.9”) long, USA made Enviro-Safe® thermometers are EnviroKleen certified environmentally friendly and features 100% non-toxic, non-hazardous, biodegradable, Enviro-Safe® green liquid against white back glass, and recyclable Safety-Pak packaging. Enviro-Safe® thermometers have the same accuracy as similar mercury thermometers, but without the hazards. Item B60510-0100 features PFA flouropolymer safety coating to keep liquid and glass encapsulated in the event of breakage.Individually serialized and hand inspected by our U.S. quality team. They are packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Enviro-Safe is a registered trademark of H-B Instrument Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service