Skip to Content
Merck
All Photos(1)

Documents

B5151

Sigma-Aldrich

Benzil

98%

Synonym(s):

Dibenzoyl, Diphenylethanedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCOC6H5
CAS Number:
Molecular Weight:
210.23
Beilstein:
608047
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

1 mmHg ( 128.4 °C)

Assay

98%

form

crystals

mp

94-95 °C (lit.)

SMILES string

O=C(c1ccccc1)C(=O)c2ccccc2

InChI

1S/C14H10O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10H

InChI key

WURBFLDFSFBTLW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

356.0 °F - closed cup

Flash Point(C)

180 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anna Hoerder-Suabedissen et al.
Cerebral cortex (New York, N.Y. : 1991), 28(5), 1882-1897 (2018-02-27)
The thalamus receives input from 3 distinct cortical layers, but input from only 2 of these has been well characterized. We therefore investigated whether the third input, derived from layer 6b, is more similar to the projections from layer 6a
Tsuneomi Kawasaki et al.
Organic letters, 10(18), 4085-4088 (2008-08-30)
Chiral crystals of achiral benzils act as efficient chiral initiators of asymmetric autocatalysis to afford highly enantioenriched pyrimidyl alkanols whose absolute configurations depend upon the enantiomorph of the crystal used in conjunction with asymmetric autocatalysis with amplification of enantiomeric excess.
Jonathan L Sessler et al.
Organic letters, 10(1), 73-75 (2007-12-07)
The isolation and characterization of an intermediate from the benzil-cyanide reaction is reported. The use of this trapping chemistry to produce a chemical indicator for the cyanide anion is described. It relies on the synthesis and reaction of a pi-extended
Céline Mousset et al.
Bioorganic & medicinal chemistry letters, 18(11), 3266-3271 (2008-05-15)
A series of benzil derivatives related to combretastatin A-4 (CA-4) have been synthesized by oxidation of diarylalkynes promoted by PdI(2) in DMSO. Using this new protocol, 14 benzils were prepared in good to excellent yields and their biological activity has
Zijia Li et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(6), 1802163-1802163 (2019-04-03)
Methoxy-functionalized triphenylamine-imidazole derivatives that can simultaneously work as hole transport materials (HTMs) and interface-modifiers are designed for high-performance and stable perovskite solar cells (PSCs). Satisfying the fundamental electrical and optical properties as HTMs of p-i-n planar PSCs, their energy levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service