Skip to Content
Merck
All Photos(1)

Key Documents

464279

Sigma-Aldrich

2-(Trifluoromethoxy)aniline

95%

Synonym(s):

α,α,α-Trifluoro-o-anisidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$55.10

$55.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$55.10

About This Item

Linear Formula:
CF3OC6H4NH2
CAS Number:
Molecular Weight:
177.12
Beilstein:
2803814
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$55.10


Check Cart for Availability

Request a Bulk Order

Assay

95%

refractive index

n20/D 1.461 (lit.)

bp

61-63 °C/15 mmHg (lit.)

density

1.301 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Nc1ccccc1OC(F)(F)F

InChI

1S/C7H6F3NO/c8-7(9,10)12-6-4-2-1-3-5(6)11/h1-4H,11H2

InChI key

ZFCOUBUSGHLCDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(Trifluoromethoxy)aniline may be used in the preparation of 2-amino-6-(trifluoromethoxy)benzothiazole (SKA-35).[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ananthakrishnan Sankaranarayanan et al.
Molecular pharmacology, 75(2), 281-295 (2008-10-29)
Small-conductance (KCa2.1-2.3) and intermediate-conductance (KCa3.1) calcium-activated K(+) channels are critically involved in modulating calcium-signaling cascades and membrane potential in both excitable and nonexcitable cells. Activators of these channels constitute useful pharmacological tools and potential new drugs for the treatment of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service