Skip to Content
Merck
All Photos(1)

Key Documents

Z511471

Sigma-Aldrich

Ultrasonic processors 500W and 750W

500W, AC/DC input 220 V AC, Schuko plug

Synonym(s):

sonicator, ultra sonicator, ultrasonic homogenizer

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$10,110.00

$10,110.00


Estimated to ship on12 September 2025FromTruganina



Select a Size

Change View
1 EA
$10,110.00

About This Item

UNSPSC Code:
41101519
NACRES:
NB.74

$10,110.00


Estimated to ship on12 September 2025FromTruganina


feature

CE compliant

manufacturer/tradename

Sonics & Materials VCX500-220V

Looking for similar products? Visit Product Comparison Guide

General description

The VCX 500 offers 500 watts of power to handle a range of liquid processing applications. This model shares the same microprocessor based operating system and programming screens as the VCX750 system.Using a variety of accessories, the system can process a wide range of sample types and volumes for many different applications.The VCX 500 includes a standard 1/2" diameter probe (50 – 250mL volumes) with replaceable tip.


  • Automatic tuning and frequency control
  • Integrated temperature controller from 1°C to 100°C
  • Stores up to 10 procedures
  • Real time display
  • Control process time from 1 second to 10 hours
  • Shipped complete with a 1/2 in. (13 mm) probe (630-0220) with replaceable tip, tool kit, and instruction manual

Applications:
Cell disruption in bacteria, spores, and tissues; accelerating catalytic reactions; extractions of serums, toxins, enzymes, and viruses from organic sources; solubilizing difficult compounds; emulsification to 0.01 mm; homogenizing liquids; sample preparation from microscopy and particle size analysis.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Surfactant-assisted dispersion of single-walled carbon nanotubes for debundling or exfoliation in dispersion procedures.

Related Content

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Questions

  1. What is the volume which can be handled in Z511471-1EA. is it available in 2 modes (500 W and 750 W). What is the quantity of fluid that can be handled and viscosity range of fluid for processing.

    1 answer
    1. This item is a 500W sonicator that includes a ½ in. probe. The probe may accommodate volumes ranging from 20 – 250 mL, depending on the sample type, viscosity, and vessel type. The 750W product option and additional probes are sold separately. There is no specified viscosity range. However, liquids that will not pour or circulate easily may be too thick to processed effectively. Typically, the maximum viscosity at which a material can be processed effectively is 4000 cps.
      Please see the link below to review the product user guide:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/296/249/sonics-userguide.pdf

      Please see the following link to review additional products and accessories that may be of interest:
      https://www.sigmaaldrich.com/search/ultrasonic-processors-500w-and-750w?focus=products&page=1&perpage=30&sort=relevance&term=Ultrasonic%20processors%20500W%20and%20750W&type=product

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service