Skip to Content
Merck
All Photos(1)

Key Documents

93528

Sigma-Aldrich

Ethyl azidoacetate solution

~25% in ethanol (NMR)

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$316.00

$316.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 ML
$316.00

About This Item

Linear Formula:
N3CH2COOCH2CH3
CAS Number:
Molecular Weight:
129.12
Beilstein:
4247209
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$316.00


Available to ship on15 April 2025Details


Request a Bulk Order

form

solution

Quality Level

reaction suitability

reaction type: click chemistry

concentration

~25% in ethanol (NMR)

functional group

azide
ester

SMILES string

CCOC(=O)CN=[N+]=[N-]

InChI

1S/C4H7N3O2/c1-2-9-4(8)3-6-7-5/h2-3H2,1H3

InChI key

HVJJYOAPXBPQQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service