Skip to Content
Merck
All Photos(1)

Key Documents

88539

Sigma-Aldrich

Ethyl azidoacetate solution

~30% in methylene chloride (NMR)

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$173.00

$173.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$173.00

About This Item

Linear Formula:
N3CH2COOCH2CH3
CAS Number:
Molecular Weight:
129.12
Beilstein:
4247209
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$173.00


Check Cart for Availability

Request a Bulk Order

form

solution

concentration

~30% in methylene chloride (NMR)

functional group

azide
ester

SMILES string

CCOC(=O)CN=[N+]=[N-]

InChI

1S/C4H7N3O2/c1-2-9-4(8)3-6-7-5/h2-3H2,1H3

InChI key

HVJJYOAPXBPQQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service