Skip to Content
Merck
All Photos(1)

Key Documents

935271

Sigma-Aldrich

Fully Equipped H-Cell Kit (50 ml Volume)

Synonym(s):

IP-HC-50

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$2,440.00

$2,440.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$2,440.00

About This Item

UNSPSC Code:
41111761
NACRES:
NA.21

$2,440.00


Check Cart for Availability

Request a Bulk Order

General description

Our Fully Equipped H-Cell Kit (50 ml Volume) is a fully equipped, horizontally mounted and a dual compartment electrochemical H-Cell set-up for a wide variety of studies that require a separate compartment for counter and / or reference electrodes. The construction is gas-tight having two separate chambers, each equipped with gas inlet and outlet. This allows bubbling the solution and evacuating gases. Chambers can be separated with an ion-exchange membrane, so the electrochemical products appearing at working and counter electrode do not affect the opposite electrode. Set-up is compatible with a power source meter or a potentiostat / Galvanostat.
It includes: Glass reservoirs with three-electrode set-up, Spacer, Grip nuts, Screws, Purge adaptor, PTFE support holder, Top silicon dummy, Plugs, Silicon tube 0.5 meters, Silicon septa.

Application

The product can be used in various applications such as:
  • Electrochemical water splitting - HER / OER or any such electrocatalysis applications
  • Redox flow battery setup
  • Carbon Dioxide Reduction Studies
  • Membrane research: Proton conductivity, Ionic conductivity, membrane efficiency
  • Reference electrode testing
  • ASTM grade Corrosion studies – Hydrogen Permeation
  • Fuel Cell Studies: Hydrogen peroxide fuel cell, enzymatic fuel cell research, microbial fuel cell research, etc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service