Skip to Content
Merck
All Photos(1)

Key Documents

933031

Sigma-Aldrich

Surface Mount Tweezers

Style 110

Synonym(s):

Tweezers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$106.00

$106.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$106.00

About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

$106.00


Check Cart for Availability

Request a Bulk Order

General description

EMS techware
Surface Mount Tweezers Style 110 are straight "T" shaped tips with a vertical groove inside for holding cylindrical objects up to 1mm diameter. Length: 4¾" (120 mm).

Application

Absolutely perfect for holding and installing Omniprobe Tips. Especially if you′re using Cryo-lift out and have to install the tip with the rod already inserted into the Scanning Electron Microscope (SEM) chamber.
Forceps provide a firm grip and longitudinal stability reducing the chance of dropping the tip.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service