Key Documents
Z162507
Sigma-Aldrich
All-purpose tweezer set
5 tweezers in a vinyl pouch, pkg of 1 set
Select a Size
1 SET
$123.00
$123.00
Available to ship on24 April 2025Details
Select a Size
Change View
1 SET
$123.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
$123.00
Available to ship on24 April 2025Details
packaging
pkg of 1 set
General description
Contains 5 tweezers in a vinyl pouch: 41/2 in. dissecting tweezer, 43/4 in. stamp tweezer, 6 in. retrieving forceps, 6 in. solder utility tweezer, and 61/2 in. self-closing tweezer.
Choose from one of the most recent versions:
Certificates of Analysis (COA)