Skip to Content
Merck
All Photos(1)

Documents

92825

Sigma-Aldrich

2,3,6-Trinitrophenol

moistened with water, ≥95.0% (calc. based on dry substance, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3N3O7
CAS Number:
Molecular Weight:
229.10
Beilstein:
3149687
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0% (calc. based on dry substance, HPLC)

quality

moistened with water

contains

~40% water as stabilizer

mp

119-120 °C (lit.)

SMILES string

Oc1c(ccc(c1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O7/c10-6-4(8(13)14)2-1-3(7(11)12)5(6)9(15)16/h1-2,10H

InChI key

UPOHJPYGIYINKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Flam. Sol. 2

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service