Skip to Content
Merck
All Photos(1)

Key Documents

928224

Sigma-Aldrich

Gold nanoparticles

70 nm diameter, azide functionalized, 1ml, OD 50, in water

Synonym(s):

AuNP, GdNP, Gold NP

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$946.00

$946.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$946.00

About This Item

UNSPSC Code:
12141717
NACRES:
NA.22

$946.00


Estimated to ship on13 April 2025


Request a Bulk Order

Quality Level

description

Alkyne surface density: 1/nm2

concentration

in water

OD

50

core diam.

70 nm , Coefficient of Variance < 12%

diameter

70 nm

λmax

548 nm

PDI

<0.200

functional group

azide

storage temp.

2-8°C

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azide functionalized gold nanoparticles are suitable for covalent conjugation of any molecule with a terminal alkyne through click chemistry. These gold nanoparticles allow for rapid generation of gold conjugates for subsequent use in downstream assays.

Azide functionalized gold nanoparticles are more than 95% spherical and have uniform size distribution (CV <12%).

Application

Ideal for development of gold conjugates for use in applications such as blotting, lateral flow assays, microscopy, and transmission electron microscopy (TEM).

Storage and Stability

When stored for a long period of time gold nanoparticles may sediment at the bottom of the vial, which is especially prominent for larger particle sizes. Prior to use, re-suspend the sedimented particles by thorough mixing until a homogenous solution is obtained.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L Brennan et al.
Bioconjugate chemistry, 17(6), 1373-1375 (2006-11-16)
A simple and versatile method for the preparation of functional enzyme-gold nanoparticle conjugates using "click" chemistry has been developed. In a copper-catalyzed 1,2,3-triazole cycloaddition, an acetylene-functionalized Thermomyces lanuginosus lipase has been attached to azide-functionalized water-soluble gold nanoparticles under retention of
Monika Fischler et al.
Chemical communications (Cambridge, England), (2), 169-171 (2007-12-20)
We present a new type of azide-functionalized gold nanoparticle and their coupling to an alkyne-modified DNA duplex using the copper(I)-catalyzed Huisgen cycloaddition ('click chemistry'), resulting in a chain-like assembly of nanoparticles on the DNA template.
Vivek Poonthiyil et al.
Beilstein journal of organic chemistry, 14, 11-24 (2018-01-31)
Glycoscience, despite its myriad of challenges, promises to unravel the causes of, potential new detection methods for, and novel therapeutic strategies against, many disease states. In the last two decades, glyco-gold nanoparticles have emerged as one of several potential new

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service