Gold nanoparticles
Synonym(s):
Au NP, Gold Colloid, Gold colloid, Gold nanoparticles NH2 functionalized, Au NP NH2, Gold/Silica core/shell particles, Gold nanoparticles COOH functionalized, Au NP COOH, Octylthiol functionalized gold nanoparticles, Au nanoparticles, Gold nanoparticles OMe terminated, Au NP OMe
Products
Brand
pH Value
Physical Form
Quality Segment
Functional Group
Package Size
Showing 1-20 of 84
Page 1 of 5
Compare
Product Number
Description
Pricing
Page 1 of 5