Skip to Content
Merck
All Photos(1)

Key Documents

927856

Sigma-Aldrich

Gold nanoparticles

10 nm diameter, azide functionalized, 1ml, OD 50, In in water

Synonym(s):

AuNP, GdNP, Gold NP

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$881.00

$881.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$881.00

About This Item

UNSPSC Code:
12141717
NACRES:
NA.23

$881.00


Check Cart for Availability

Request a Bulk Order

Quality Level

description

Azide surface density: 1/nm2

concentration

In in water

OD

50

core diam.

10 nm , Coefficient of variance <12%

diameter

10 nm

λmax

520 nm

PDI

<0.200

functional group

azide

storage temp.

2-8°C

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azide functionalized gold nanoparticles are suitable for covalent conjugation of any molecule with a terminal alkyne through click chemistry. These gold nanoparticles allow for rapid generation of gold conjugates for subsequent use in downstream assays.

Azide functionalized gold nanoparticles are more than 95% spherical and have uniform size distribution (CV <12%).

Application

Ideal for development of gold conjugates for use in applications such as blotting, lateral flow assays, microscopy, and transmission electron microscopy (TEM).

Storage and Stability

When stored for a long period of time gold nanoparticles may sediment at the bottom of the vial, which is especially prominent for larger particle sizes. Prior to use, re-suspend the sedimented particles by thorough mixing until a homogenous solution is obtained.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent applications of click chemistry for the functionalization of gold nanoparticles and their conversion to glyco-gold nanoparticles
Poonthiyil, Vivek, et al
Beilstein Journal of Organic Chemistry, 14.1, 11-24 (2018)
Chain-like assembly of gold nanoparticles on artificial DNA templates via `click chemistry?
Fischler, Monika, et al
Chemical Communications (Cambridge, England), 2, 169-171 (2008)
Bionanoconjugation via click chemistry: The creation of functional hybrids of lipases and gold nanoparticles
Brennan, Jennifer L., et al
Bioconjugate Chemistry, 17.6, 1373-1375 (2006)

Articles

Explore DNA-functionalized nanoparticles in sensors for precise target analyte detection. Learn about types, synthesis, functionalization, and design optimization, with insights into challenges and prospects.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service