Skip to Content
Merck
All Photos(2)

Key Documents

923524

Sigma-Aldrich

(tBuMPhos)PdCl2

greener alternative

Synonym(s):

1-Diadamantylphosphino-1′-di-tert-butylphosphinoferrocene palladium dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$121.00
500 MG
$364.00

$121.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$121.00
500 MG
$364.00

About This Item

Empirical Formula (Hill Notation):
C38H56Cl2FeP2Pd
Molecular Weight:
807.97
UNSPSC Code:
12352200
NACRES:
NA.21

$121.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[Cl-][Pd+2]1([Cl-])[P](C(C)(C)C)(C(C)(C)C)[C-]23[CH]4=[CH]5[CH]6=[CH]2[Fe+2]56789%1043[CH]=%11[CH]%10=[CH]9[C-]8([CH]%117)[P]1(C%12%13CC%14CC(CC(C%14)C%12)C%13)C%15%16CC%17CC(CC(C%17)C%15)C%16

InChI

InChI=1S/C25H34P.C13H22P.2ClH.Fe.Pd/c1-2-4-23(3-1)26(24-11-17-5-18(12-24)7-19(6-17)13-24)25-14-20-8-21(15-25)10-22(9-20)16-25;1-12(2,3)14(13(4,5)6)11-9-7-8-10-11;;;;/h1-4,17-22H,5-16H2;7-10H,1-6H3;2*1H;;/q2*-1;;;2*+2/p-2

InChI key

BJILVUSTVVKBGK-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

(tBuMPhos)PdCl2 is an unsymmetrical ferrocene-based phosphine ligand. The MPhos series of ligands are unsymmetrical diphosphino-ferrocenes bearing a bulky adamantyl group. Pd catalysts containing CyMPhos can be used for a variety of cross-coupling transformations, including Suzuki coupling, C-P coupling, and Csp2-Csp3 coupling. Demonstrated Csp2-Csp3 couplings include Murahashi-Feringa Coupling (Alkyl-Li), Kumada-Corriu coupling (Alkyl-MgX), Negishi coupling (Alkyl-ZnX) and Suzuki–Miyaura coupling (Alkyl-B(OR2)).

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service