Skip to Content
Merck
All Photos(2)

Key Documents

923443

Sigma-Aldrich

PhMPhos

greener alternative

Synonym(s):

1-Diadamantylphosphino-1′-diphenylphosphinoferrocene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$117.00
500 MG
$348.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$117.00
500 MG
$348.00

About This Item

Empirical Formula (Hill Notation):
C42H48FeP2
Molecular Weight:
670.62
UNSPSC Code:
12352200
NACRES:
NA.21

$117.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

functional group

phosphine

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

PhMPhos is an unsymmetrical ferrocene-based phosphine ligand. The MPhos series of ligands are unsymmetrical diphosphino-ferrocenes bearing a bulky adamantyl group. Pd catalysts containing PhMPhos can be used for a variety of cross-coupling transformations, including Suzuki coupling, C-P coupling, and Csp2-Csp3 coupling. Demonstrated Csp2-Csp3 couplings include Murahashi-Feringa Coupling (Alkyl-Li), Kumada-Corriu coupling (Alkyl-MgX), Negishi coupling (Alkyl-ZnX) and Suzuki–Miyaura coupling (Alkyl-B(OR2)).

Learn more about MPhos ligands and catalysts

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service