Skip to Content
Merck
All Photos(3)

Key Documents

677264

Sigma-Aldrich

APhos

95%

Synonym(s):

(4-(N,N-Dimethylamino)phenyl)di-tert-butyl phosphine, A-taPhos, [4-(Dimethylamino)phenyl]bis(tert-butyl)phosphine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$101.00
5 G
$254.00

$101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$101.00
5 G
$254.00

About This Item

Empirical Formula (Hill Notation):
C16H28NP
CAS Number:
Molecular Weight:
265.37
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$101.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reagent type: ligand
reaction type: Cross Couplings

mp

57-61 °C

functional group

phosphine

SMILES string

CN(C)c1ccc(cc1)P(C(C)(C)C)C(C)(C)C

InChI

1S/C16H28NP/c1-15(2,3)18(16(4,5)6)14-11-9-13(10-12-14)17(7)8/h9-12H,1-8H3

InChI key

IQTHEAQKKVAXGV-UHFFFAOYSA-N

Application

Ligand used to prepare a palladium dichloride catalyst (678740) on treatment with PdCl2(COD). The catalyst effectively cross-couples aryl boronic acids with heteroaryl chlorides.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anil S Guram et al.
Organic letters, 8(9), 1787-1789 (2006-04-21)
[reaction: see text] New air-stable PdCl(2){P(t)Bu(2)(p-R-Ph)}(2) (R = H, NMe(2), CF(3),) complexes represent simple, general, and efficient catalysts for the Suzuki-Miyaura cross-coupling reactions of aryl halides including five-membered heteroaryl halides and heteroatom-substituted six-membered heteroaryl chlorides with a diverse range of

Articles

Ligand used to prepare a palladium dichloride catalyst on treatment with PdCl2(COD). The catalyst effectively cross-couples aryl boronic acids with heteroaryl chlorides.

Related Content

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service