Skip to Content
Merck
All Photos(1)

Key Documents

921602

Sigma-Aldrich

Nickel oxide nanoparticle ink

2.5 wt. %, viscosity 1.3 cP 

Synonym(s):

NiO dispersion, NiO ink, Nickel oxide dispersion

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$175.00

$175.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$175.00

About This Item

Empirical Formula (Hill Notation):
NiO
CAS Number:
Molecular Weight:
74.69
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

$175.00


Check Cart for Availability

Request a Bulk Order

form

suspension

Quality Level

concentration

2.5 wt. %

viscosity

1.3 cP

density

0.8 g/mL at 25 °C

SMILES string

[Ni]=O

InChI

1S/Ni.O

InChI key

GNRSAWUEBMWBQH-UHFFFAOYSA-N

General description

This nickel oxide nanoparticle ink is a nanoparticle based printing ink allowing processing temperatures of 80-120°C. The nickel oxide nanoparticle ink is a hole selective interface layer ink containing nickel oxide (NiO) nanoparticles in ethanol. The NiO nanoparticle ink functions well as hole extraction layer (HEL) material in solar cells.

Application

This NiO nanoparticle ink is for slot-dye, spin-coating and doctor blading for the use as hole transport layer in printed electronics. The NiO nanoparticle ink is universally applicable in normal and inverted architecture.

Preparation Note

  • Storage: In dark at room temperature.
  • Prior to application: Shake, ultrasonicate with sonic horn (50W, 1min/100mL) or bath (5min/100mL). Optionally filter through 0.45μm PP or PTFE filter.
  • Post-treatment: Annealing of deposited films at 80°C - 120°C.
  • Thickness optimization may be necessary.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

56.3 °F

Flash Point(C)

13.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find advantages of inorganic interface layer inks for organic electronic & other applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service