Skip to Content
Merck
All Photos(1)

Key Documents

901143

Sigma-Aldrich

PDI-DPP-PDI

Synonym(s):

Perylene diimide flanked diketopyrrolopyrrole, nPDI2DPP

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$623.00

$623.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$623.00

About This Item

Empirical Formula (Hill Notation):
C110H118N8O10S2
CAS Number:
Molecular Weight:
1776.29
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

$623.00


Check Cart for Availability

Request a Bulk Order

description

Band gap: 1.6 eV

Quality Level

form

powder

Orbital energy

HOMO -5.3 eV 
LUMO -3.7 eV 

SMILES string

O=C1C(C2=C(C3=CC=C(C4=CC(C(N(C(CC)CC)C5=O)=O)=C(C5=CC(N6CCCCCC)=C78)C7=C4C9=CC=C%10C%11=C(C(N(C(CC)CC)C%10=O)=O)C=C6C8=C9%11)S3)N1CCCCCCCC)=C(C%12=CC=C(C%13=CC(C(N(C(CC)CC)C%14=O)=O)=C(C%14=CC(N%15CCCCCC)=C%16%17)C%16=C%13C%18=CC=C%19C%20=C(C(N(C(CC)CC)C%

Legal Information

Sold under Material Transfer Agreement with Gregory Welch group at University of Calgary.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simply Complex: The Efficient Synthesis of an Intricate Molecular Acceptor for High-Performance Air-Processed and Air-Tested Fullerene-Free Organic Solar Cells.
McAfee SM, et al.
Chemistry of Materials, 29(3), 1309-1314 (2017)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service