Skip to Content
Merck
All Photos(1)

Key Documents

920371

Sigma-Aldrich

Lithium hexafluorophosphate

99.9% trace metals basis

Synonym(s):

LBG 45864, Lithium fluophosphate, Lithium phosphorus fluoride

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$467.00
250 G
$1,770.00

$467.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$467.00
250 G
$1,770.00

About This Item

Linear Formula:
LiPF6
CAS Number:
Molecular Weight:
151.91
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
for analytical purposes

$467.00


Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

99.9% trace metals basis

mp

200 °C (dec.) (lit.)

SMILES string

[Li+].F[P-](F)(F)(F)(F)F

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1

InChI key

AXPLOJNSKRXQPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hexafluorophosphate 99.9% trace metals basis is an inorganic compound that comes in white color powder. It is highly soluble in organic solvents, such as dimethyl carbonate (DMC) and ethylene carbonate (EC). It possesses excellent ionic conductivity and electrochemical stability.

Application

Lithium hexafluorophosphate (LiPF6) 99.9% trace metals basis is primarily designed for R&D activities where it can be used as an electrolyte salt in lithium-ion batteries, enhancing ionic conductivity and electrochemical stability. Its high purity (99.9% trace metals basis) ensures minimal impurities, which is essential for preserving the performance and lifespan of the batteries. It can be used in research focused on the formation and stability of the SEI layer on anode surfaces. LiPF6 is used in the electrolyte solutions of supercapacitors to enhance their energy density and cycle life. It can be used to design electrolytes for improving the performance of energy storage devices.[1][2][3]

Features and Benefits

  • 99.9% of trace metals ensure minimal impurities, which is essential for accurate and reproducible results.
  • Lower amount of trace metals Ca, Cr, Mg and K ensures performance

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 1 Inhalation

Target Organs

Bone,Teeth

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jung-Gu Han et al.
Advanced materials (Deerfield Beach, Fla.), 31(20), e1804822-e1804822 (2018-11-13)
In conjunction with electrolyte additives used for tuning the interfacial structures of electrodes, functional materials that eliminate or deactivate reactive substances generated by the degradation of LiPF6 -containing electrolytes in lithium-ion batteries offer a wide range of electrolyte formulation opportunities.
Preparation and characterization of lithium hexafluorophosphate for lithium-ion battery electrolyte.
Liu J, et al.
Transactions of Nonferrous Metals Society of China, 20(2), 344-348 (2010)
Preparation and characterization of lithium hexafluorophosphate for lithium-ion battery electrolyte
Liu J, et al.
Transactions of Nonferrous Metals Society of China, 20(2), 344-348 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service