Skip to Content
Merck
All Photos(2)

Documents

916080

Sigma-Aldrich

Diethyl-4-cyclohexyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate

≥95%

Synonym(s):

4-Alkyl-1,4-dihydropyridine reagent, DHP reagent for light-mediated His modification

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H29NO4
CAS Number:
Molecular Weight:
335.44
MDL number:
UNSPSC Code:
12352200

Assay

≥95%

form

powder

storage temp.

−20°C

InChI

1S/C19H29NO4/c1-5-23-18(21)15-12(3)20-13(4)16(19(22)24-6-2)17(15)14-10-8-7-9-11-14/h14,17,20H,5-11H2,1-4H3

InChI key

GERWBKSVDHUVIT-UHFFFAOYSA-N

Application

Diethyl-4-cyclohexyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate was reported to chemoselectively modify histidine under visible light where the unsubstituted nitrogen groups on the modified His imidazole are conserved. Diethyl-4-cyclohexyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate is also a versitile reagent for photoredox chemistry.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuefeng Wang et al.
Chemical communications (Cambridge, England), 55(43), 6010-6013 (2019-05-08)
A sulfonylation reaction of 4-substituted Hantzsch esters, DABCO·(SO2)2, and electron-deficient alkenes at room temperature in the presence of photoredox catalysis under visible light irradiation is described. Not only (E)-chalcones but also (vinylsulfonyl)benzene and 2-vinylpyridine are all suitable substrates in the
Kaiqian Wang et al.
Organic & biomolecular chemistry, 17(15), 3845-3852 (2019-04-03)
Herein, we report a simple, economical, and effective acid-mediated method for the in situ deuteration of Hantzsch esters and their 4-substituted derivatives, including some drugs that constitute important calcium channel blockers which are effective for hypertension treatment. Hydrogen isotope exchange
Jennifer K Matsui et al.
Angewandte Chemie (International ed. in English), 57(48), 15847-15851 (2018-10-12)
A regioselective, nickel-catalyzed photoredox allylation of secondary, benzyl, and α-alkoxy radical precursors is disclosed. Through this manifold, a variety of linear allylic alcohols and allylated monosaccharides are accessible in high yields under mild reaction conditions. Quantum mechanical calculations [DFT and
Hai-Wu Du et al.
Organic letters, 22(4), 1542-1546 (2020-01-29)
In this study, a facile and efficient method to synthesize monofluoroalkenes by photoredox catalytic defluorinative alkylation of gem-difluoroalkenes with 4-alkyl-1,4-dihydropyridines under mild conditions (room temperature) is described. This novel strategy is applicable for a broad range of gem-difluoroalkene substrates with
Kazunari Nakajima et al.
Organic letters, 21(12), 4642-4645 (2019-05-31)
Introduction of alkyl groups on azodicarboxylate esters is an important method to prepare alkyl amine derivatives. Herein, we report reactions of 4-alkyl-1,4-dihydropyridines as alkylation reagents with di- tert-butyl azodicarboxylate to prepare alkyl amine derivatives under heating conditions. The alkylation reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service