Skip to Content
Merck
All Photos(1)

Key Documents

914746

Sigma-Aldrich

Thiophosphoro alkyne dichloridate

≥95%

Synonym(s):

O-(Prop-2-yn-1-yl) phosphorodichloridothioate, Chemoselective histidine labeling reagent, Phosphorodichloridothioate 2-propyn-1-ol, Propargyl thiophosphorodichloridate, TPAC, Thiophosphoro alkyne dichloridate, Thiophosphorodichloridate reagent, Thiophosphorus electrophile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$598.00

$598.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$598.00

About This Item

Empirical Formula (Hill Notation):
C3H3Cl2OPS
CAS Number:
Molecular Weight:
189.00
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

$598.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

liquid

storage temp.

−20°C

SMILES string

[P](=S)(Cl)(Cl)OCC#C

InChI

1S/C3H3Cl2OPS/c1-2-3-6-7(4,5)8/h1H,3H2

InChI key

HPJLQEFYLAALCL-UHFFFAOYSA-N

Application

Thiophosphoro alkyne dichloridate (TPAC) is a histidine labeling reagent developed by the Chang lab that mimics native post-translational His phosphorylation. The alkyne allows for subsequent reactions or addition of payloads by copper-assisted alkyne-azide cycloaddition (CuAAC) chemistry. TPAC was also demonstrated to covalently modify His-tags, and by clicking on polyarginine tags, enabled protein delivery into live cells.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

175.8 °F

Flash Point(C)

79.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shang Jia et al.
Journal of the American Chemical Society, 141(18), 7294-7301 (2019-04-25)
Site-selective bioconjugation to native protein residues is a powerful tool for protein functionalization, with cysteine and lysine side chains being the most common points for attachment owing to their high nucleophilicity. We now report a strategy for histidine modification using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service