Skip to Content
Merck
All Photos(1)

Key Documents

913960

Sigma-Aldrich

Conjugated Polymer Nanoparticles incorporating iron oxide

fluorescence λem 420 nm, 100 μg/mL in H2O

Synonym(s):

Conjugated polymer nanoparticles, Fluorescent magnetic nanoparticles, Fluorescent nanoparticles, Magneto-conjugated polymer nanoparticles

Sign Into View Organizational & Contract Pricing

Select a Size

250 μL
$819.00

$819.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 μL
$819.00

About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

$819.00


Check Cart for Availability

Request a Bulk Order

form

nanoparticles

Quality Level

concentration

100 μg/mL in H2O

fluorescence

λem 420 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Iron oxide containing conjugated polymer nanoparticles are water-soluble nanoparticles compromising of light-emitting polymer and iron oxide. The presence of iron oxide in core makes these fluorescent nanoparticles distinctly different than conventional dyes as it allows their manipulation with a magnetic field. These nanoparticles possess a ′core-shell′ structure where light-emitting polymer and iron oxide are encapsulated in biocompatible surfactant.[1][2]
The ′core-shell′ structure consisting of polymer and iron oxide in the core and the biocompatible surfactant in the surrounding shell allows easy covalently bonding of these nanoparticles with biomolecules such streptavidin, antibodies, proteins, nucleic acids, etc and make them suitable for the following applications:
  • Fluorescence Microscopy (high content screening, live cell tracking and 3-D cell imaging)
  • Immunohistochemistry and Immunocytochemistry
  • Flow Cytometry
  • Rapid Diagnostic Tests (RDTs)
  • Tissue Imaging
  • 2-Photon Imaging
  • NIR Imaging

Storage and Stability

The robustness of CPNs enables them to be used under high intensity illumination and in extreme experimental conditions if required, while their 24-month shelf-life permits convenient storage at ambient temperatures

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General synthesis of high-performing magneto-conjugated polymer core?shell nanoparticles for multifunctional theranostics.
Yan H, et al.
Nano Research, 10, 704-704 (2017)
Philip Howes et al.
Journal of the American Chemical Society, 132(28), 9833-9842 (2010-06-25)
Hybrid nanoparticles which incorporate multiple functionalities, such as fluorescence and magnetism, can exhibit enhanced efficiency and versatility by performing several tasks in parallel. In this study, magnetic-fluorescent semiconductor polymer nanospheres (MF-SPNs) have been synthesized by encapsulation of hydrophobic conjugated polymers

Articles

CPNs offer sensitivity and stability, aiding various assays; may enable rapid diagnostic tests for healthcare needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service