Skip to Content
Merck
All Photos(1)

Key Documents

912255

Sigma-Aldrich

sBOX(tBu)

≥95%

Synonym(s):

Di-tert-butyl 2,2′-(cyclopropane-1,1-diyl)(4S,4′S)-bis(4,5-dihydrooxazole-4-carboxylate)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$471.00

$471.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$471.00

About This Item

Empirical Formula (Hill Notation):
C19H28N2O6
CAS Number:
Molecular Weight:
380.44
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

$471.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder or solid

mp

120.5 °C

storage temp.

2-8°C

SMILES string

N1=C(OC[C@H]1C(=O)OC(C)(C)C)C3(CC3)C2=N[C@@H](CO2)C(=O)OC(C)(C)C

InChI

1S/C19H28N2O6/c1-17(2,3)26-13(22)11-9-24-15(20-11)19(7-8-19)16-21-12(10-25-16)14(23)27-18(4,5)6/h11-12H,7-10H2,1-6H3/t11-,12-/m0/s1

InChI key

LKXTWWHADOFMCC-RYUDHWBXSA-N

Application

sBOX(tBu) is a serine-derived chiral bisoxazoline ligand. The additional coordinating functionality provided by the esters enables new reactivity compared to traditional BOX ligands. sBOX(iPr) has been shown to be an optimal ligand for the electrocatalytic cyanophosphorylation as well as hydrocyanation of vinylarenes.

Product can be used with our Synlectro electrochemistry platform (ESYNTH001)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niankai Fu et al.
Journal of the American Chemical Society, 141(37), 14480-14485 (2019-09-10)
In contrast to the rapid growth of synthetic electrochemistry in recent years, enantioselective catalytic methods powered by electricity remain rare. In this work, we report the development of a highly enantioselective method for the electrochemical cyanophosphinoylation of vinylarenes. A new

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service