Skip to Content
Merck
All Photos(2)

Documents

692778

Sigma-Aldrich

(R,R)-DACH-naphthyl Trost ligand

95%

Synonym(s):

(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N′-bis(2-diphenylphosphino-1-naphthoyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H44N2O2P2
CAS Number:
Molecular Weight:
790.87
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

optical activity

[α]20/D +69.0°, c = 1 in methanol

mp

224-231 °C

SMILES string

O=C(N[C@@H]1CCCC[C@H]1NC(=O)c2c(ccc3ccccc23)P(c4ccccc4)c5ccccc5)c6c(ccc7ccccc67)P(c8ccccc8)c9ccccc9

InChI

1S/C52H44N2O2P2/c55-51(49-43-29-15-13-19-37(43)33-35-47(49)57(39-21-5-1-6-22-39)40-23-7-2-8-24-40)53-45-31-17-18-32-46(45)54-52(56)50-44-30-16-14-20-38(44)34-36-48(50)58(41-25-9-3-10-26-41)42-27-11-4-12-28-42/h1-16,19-30,33-36,45-46H,17-18,31-32H2,(H,53,55)(H,54,56)/t45-,46-/m1/s1

InChI key

VXFKMKXTPXVEMU-AWSIMMLFSA-N

Application

Asymmetric allylic alkylation ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trost, B.M. et al.
Aldrichimica Acta, 40, 59-59 (2007)
Asymmetric Transition Metal-Catalyzed Allylic Alkylations.
Barry M. Trost et al.
Chemical reviews, 96(1), 395-422 (1996-02-01)
Trost, B.M. et al.
Journal of the American Chemical Society, 124, 11616-11616 (2004)
Asymmetric transition-metal-catalyzed allylic alkylations: applications in total synthesis.
Barry M Trost et al.
Chemical reviews, 103(8), 2921-2944 (2003-08-14)

Articles

Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service