Skip to Content
Merck
All Photos(2)

Documents

909661

Sigma-Aldrich

d0-DMDSSO crosslinker

≥95%

Synonym(s):

Bis(2,5-dioxopyrrolidin-1-yl) 3,3′-sulfinylbis(2-methylpropanoate), Mass spectrometry-cleavable crosslinker for studying protein-protein interactions

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O9S
CAS Number:
Molecular Weight:
416.40
UNSPSC Code:
12352106

Assay

≥95%

form

powder

storage temp.

2-8°C

Application

d0-DMDSSO (dimethyl-disuccinimidyl sulfoxide) crosslinker is a homobifunctional, lysine residue-targeting, sulfoxide-containing crosslinker for analysis of protein-protein interactions (PPIs) through crosslinking mass spectrometry (XL-MS). d0-DMDSSO possesses two N-hydroxysuccinimide (NHS) ester groups for targeting amines and two symmetrical C-S cleavable bonds adjacent to the central sulfoxide. The post-cleavage spacer yields tagged peptides for unambiguous identification by collision-induced dissociation in tandem MS. d0-DMDSSO crosslinker will also be a useful proteomics tool and aid in the quest for targeting ″undruggable″ protein targets.

Legal Information

Subject to US Patent Application #15/275,001 of the Regents of the University of California

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinton Yu et al.
Analytical chemistry, 86(4), 2099-2106 (2014-01-30)
Structural characterization of protein complexes is essential for the understanding of their function and regulation. However, it remains challenging due to limitations in existing tools. With recent technological improvements, cross-linking mass spectrometry (XL-MS) has become a powerful strategy to define

Articles

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service