Skip to Content
Merck
All Photos(1)

Key Documents

CF0041

Sigma-Aldrich

Urea crosslinker – C4-arm, NHS ester (DSBU, BuUrBU)

Synonym(s):

1,1′-Bis(2,5-dioxo-1-pyrrolidinyl) 4,4′-(carbonyldiimino)bis[butanoate], Disuccinimidyldibutyric urea

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$1,180.00

$1,180.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 MG
$1,180.00

About This Item

Linear Formula:
C17H22N4O9
CAS Number:
MDL number:
UNSPSC Code:
12161502
NACRES:
NA.22

$1,180.00


Available to ship on22 April 2025Details


Request a Bulk Order

form

solid

reaction suitability

reaction type: C-C Bond Formation
reagent type: cross-linking reagent

SMILES string

N2(C(=O)CCC2=O)OC(=O)CCCNC(=O)NCCCC(=O)ON1C(=O)CCC1=O

InChI

1S/C17H22N4O9/c22-11-5-6-12(23)20(11)29-15(26)3-1-9-18-17(28)19-10-2-4-16(27)30-21-13(24)7-8-14(21)25/h1-10H2,(H2,18,19,28)

InChI key

XZSQCCZQFXUQCY-UHFFFAOYSA-N

Application

The ureido-4,4´-dibutyric acid bis(hydroxysuccinimide) ester, also known as DSBU or BuUrBU, is the longest version of MS-cleavable urea-based lysine-lysine reactive homobifunctional cross-linkers, featuring a C4-arm. At neutral or slightly basic pH, it irreversibly crosslinks the neighbouring lysine groups. The presence of the symmetrical urea moiety which is prone to collision-induced dissociation allows for the unambiguous distinguishing of crosslinks in tandem CID-MS experiments.

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service