Skip to Content
Merck
All Photos(3)

Key Documents

905542

Sigma-Aldrich

(R,R)-SINpEt⋅HBF4

≥95%

Synonym(s):

1,3-Bis((R)-1-(naphthalen-1-yl)ethyl)-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27BF4N2
CAS Number:
Molecular Weight:
466.32
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: ligand

functional group

phosphine

storage temp.

2-8°C

SMILES string

[B-](F)(F)(F)F.[N+]3(=CN(CC3)[C@H](C)c4c5c(ccc4)cccc5)[C@H](C)c1c2c(ccc1)cccc2

InChI key

VYIDGCVFAXSDII-MUCZFFFMSA-N

Application

(R,R)-SINpEt⋅HBF4 is a chiral NHC ligand developed in the Glorius Lab for use in Ru(II) catalyzed enantioselective hydrogenations. It has been employed for the regioselectivity asymmetric hydrogenation of quinoxalines, benzofurans, thiophenes, indolizines, isocoumarins and many other heterocycles.
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligand-controlled highly regioselective and asymmetric hydrogenation of quinoxalines catalyzed by ruthenium N-heterocyclic carbene complexes.
Slawomir Urban et al.
Angewandte Chemie (International ed. in English), 50(16), 3803-3806 (2011-03-29)
Wei Li et al.
Chemical science, 9(29), 6260-6263 (2018-08-10)
An efficient synthesis of optically active 4-substituted 2-oxazolidinones by the ruthenium(ii)-NHC-catalysed asymmetric hydrogenation of 2-oxazolones was developed. Excellent enantioselectivities (up to 96% ee) and yields (up to 99%) were obtained for a variety of substrates, bearing a range of functional
Wei Li et al.
Journal of the American Chemical Society, 139(7), 2585-2588 (2017-02-02)
A novel and practical chiral ruthenium-NHC-diamine system is disclosed for the enantioselective hydrogenation of isocoumarins, which provides a new concept to apply (chiral) NHC ligands in asymmetric catalysis. A variety of optically active 3-substituted 3,4-dihydroisocoumarins were obtained in excellent enantioselectivities

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service