Skip to Content
Merck
All Photos(2)

Key Documents

806455

Sigma-Aldrich

2,6-Dimethoxyanilino(oxo)acetic acid

Synonym(s):

[(2,6-Dimethoxyphenyl)amino](oxo)acetic acid, [(2,6-Dimethoxyphenyl)carbamoyl]formic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$372.00

$372.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$372.00

About This Item

Empirical Formula (Hill Notation):
C10H11NO5
CAS Number:
Molecular Weight:
225.20
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

$372.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: ligand

SMILES string

O=C(C(O)=O)NC1=C(OC)C=CC=C1OC

InChI

1S/C10H11NO5/c1-15-6-4-3-5-7(16-2)8(6)11-9(12)10(13)14/h3-5H,1-2H3,(H,11,12)(H,13,14)

InChI key

AULRRTNGXKPSPX-UHFFFAOYSA-N

General description

2,6-Dimethoxyanilino(oxo)acetic acid is also referred to as [(2,6-dimethylphenyl)carbamoyl]formic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Zhang et al.
Organic letters, 14(12), 3056-3059 (2012-06-06)
DMPAO has been found to be a powerful ligand to enable copper-catalyzed coupling of aryl halides with aliphatic acyclic secondary amines take place under relatively mild conditions, and coupling of aryl halides with primary amines and cyclic secondary amines proceeds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service