Skip to Content
Merck
All Photos(1)

Key Documents

901975

Sigma-Aldrich

SunTronic® silver nanoparticle ink for inkjet printing

sintering temperature 100 - 150°C

Synonym(s):

Ag ink, Conductive ink, Silver ink, SunTronic®EMD5800

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag
Molecular Weight:
107.87
UNSPSC Code:
12352200
NACRES:
NA.23

description

Silver content: 45-50%
Sintering temperature: 100 - 150°C for 10 – 60 min
Solid content: 48-52%
Volume Resistivity: 5 – 25 uΩcm

form

liquid

surface tension

25-28 dyn/cm

viscosity

6-8 cP (Brookfield)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

SunTronic® Nanosilver EMD5800 series is formulated for non-contact printing of conductive traces onto a variety of substrates.
EMD5800 is oil-based nanoparticle inks which demonstrates superior conductivity at low sintering temperatures with stable jetting performance and adhesion to temperature sensitive plastics.
It can be used in the manufacture of touch panel displays, printed RFID antennae, smart cards, sensors, security labels, touch switches and other printed electronics applications.

Caution

Printing
EMD5800 is supplied as ready-to-use oil based inks, and can be printed on various deposition printers and jetted on through various piezo DOD inkjet printheads suitable for oil based inks, although full printhead compatibility will need to be verified in each case
Sintering
Sintering conditions may change for different ink film thicknesses. Typical sintering temperature may range from 100-200°C depending on temperature tolerance of the substrate. Sintering time may range from 10-60 minutes. Electrical resistance may be reduced with higher sintering temperature and/or longer sintering times.
Storage
Refrigerated storage at a temperature of 5-15°C is recommended. Agitation after refrigeration to assure proper dispersion is recommended.

Legal Information

SunTronic is a registered trademark of Sun Chemical, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Gastrointestinal tract,Liver,Immune system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F

Flash Point(C)

69 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service