Skip to Content
Merck
All Photos(1)

Key Documents

901800

Sigma-Aldrich

Fluorescent nanodiamond

nitrogen vacancy ~3 ppm NV centers, 140 nm avg. part. size, carboxylic acid functionalized, PEG 3000 coated, 1 mg/mL in deionized water

Synonym(s):

Fluorescent nanoparticles, Nanodiamonds

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$778.00

$778.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$778.00

About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

$778.00


Check Cart for Availability

Request a Bulk Order

form

nanoparticles

composition

nitrogen vacancy, ~3 ppm NV centers

concentration

1 mg/mL in deionized water

avg. part. size

140 nm

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescent nanodiamond (nitrogen vacancy ~3 ppm NV centers, 140 nm avg. part. size, carboxylic acid functionalized, PEG 3000 coated, 1 mg/mL in deionized water), a biocompatible carbon-based nanomaterial, has interesting properties like excellent photostability, low toxic and facile surface functionalizability. It can be used for a variety of biological applications.[1][2][3]

Application

Nanodiamonds (NDs) containing NV color centres exhibit fluorescence in red/near infra-red (NIR) region. The fluorescence in NDs originates from nitrogen vacancies and, unlike quantum dots and organic dyes, NDs do not photobleach. Fluorescent NDs find applications in background-free imaging, flow cytometry, super-resolution imaging etc.[4][5][6][7][8] Our PEG coated carboxyl functionalized NDs are well suited for attachment of biomolecules using EDC chemistry. The presence PEG spacer on NDs reduces the steric hindrance among the biomolecules.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanodiamond-mediated drug delivery and imaging: Challenges and opportunities
V. Vaijayanthimala et al.,
Expert Opinion on Drug Delivery, 12, 1-1 (2014)
Nanodiamond bioconjugate probes and their collection by electrophoresis
S. C. Hens et al.,
Diamond and Related Materials, 7, 1858-1858 (2008)
Functionality is Key: Recent Progress in the Surface Modification of Nanodiamond
A. Krueger and D. Lang
Advances in Functional Materials, 22, 890-890 (2012)
Highly Fluorescent Nanodiamonds Protein-Functionalized for Cell Labeling and Targeting
B. Chang et al.,
Advances in Functional Materials, 23, 5737-5737 (2013)
The long-term stability and biocompatibility of fluorescent nanodiamond as an in vivo contrast agent
Vaijayanthimala V, et al.
Biomaterials, 33(31), 7794-7802 (2012)

Articles

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service